Thermo Scientific Chemicals

n-Docosane, 99%, Thermo Scientific Chemicals

Catalog number: A18050.14
25 g, Each
Thermo Scientific Chemicals

n-Docosane, 99%, Thermo Scientific Chemicals

Catalog number: A18050.14
25 g, Each
Quantity
Catalog number: A18050.14
also known as A18050-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-97-0
IUPAC Name
docosane
Molecular Formula
C22H46
InChI Key
HOWGUJZVBDQJKV-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCC
Assay from Supplier's CofA
≥98.5% (US-Sourced Material)
Comment
Material Sourced in UK and US
Identification (FTIR)
Conforms (UK-Sourced Material)
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid or crystalline flakes

Description

It is employed as a perfuming agent in cosmetics industry. It is also used in organic synthesis, calibration, and temperature sensing equipment. Docosane is used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a perfuming agent in cosmetics industry. It is also used in organic synthesis, calibration, and temperature sensing equipment. Docosane is used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text