Thermo Scientific Chemicals

Methyl 4-hydroxy-3,5-dimethoxybenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A18054.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-hydroxy-3,5-dimethoxybenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A18054.14
25 g, Each
Quantity
Catalog number: A18054.14
also known as A18054-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
884-35-5
IUPAC Name
methyl 4-hydroxy-3,5-dimethoxybenzoate
Molecular Formula
C10H12O5
InChI Key
ZMXJAEGJWHJMGX-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(OC)=C(O)C(OC)=C1
Appearance (Color)
Cream or pale grey
Form
Powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
102.0-108.0?C

Description

The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The scheme of laccase-catalysed iodide oxidation includes stadium of MS interaction with oxidized laccase with concomitant production of MS(ox). Thereaction of MS(ox) with iodide produced triiodide.

Solubility
Soluble in alcohol, ether, slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text