Thermo Scientific Chemicals

N-Methylbenzamide, 99%, Thermo Scientific Chemicals

Catalog number: A18057.06
5 g, Each
Thermo Scientific Chemicals

N-Methylbenzamide, 99%, Thermo Scientific Chemicals

Catalog number: A18057.06
5 g, Each
Quantity
Catalog number: A18057.06
also known as A18057-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
613-93-4
IUPAC Name
N-methylbenzamide
Molecular Formula
C8H9NO
InChI Key
NCCHARWOCKOHIH-UHFFFAOYSA-N
SMILES
CNC(=O)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
77.0-83.0?C
Form
Crystals or powder or crystalline powder

Description

It acts as a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor. It is also employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text