Thermo Scientific Chemicals

2-Bromo-2-butene, cis + trans, 98%, stab., Thermo Scientific Chemicals

Catalog number: A18058.22
100 g, Each
Thermo Scientific Chemicals

2-Bromo-2-butene, cis + trans, 98%, stab., Thermo Scientific Chemicals

Catalog number: A18058.22
100 g, Each
Quantity
Catalog number: A18058.22
also known as A18058-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13294-71-8
IUPAC Name
(2E)-2-bromobut-2-ene
Molecular Formula
C4H7Br
InChI Key
UILZQFGKPHAAOU-ONEGZZNKSA-N
SMILES
C\C=C(/C)Br
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4585-1.4635 @ 20?C

Description

Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.

Solubility
Not miscible or difficult to mix in water (664 mg/L 25 EST).

Notes
Store away from oxidizing agents and heat. Store away from electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text