Thermo Scientific Chemicals

Phosphoric acid, 85% aq. soln., Thermo Scientific Chemicals

Catalog number: A18067.0D
10,000 mL, Each
Thermo Scientific Chemicals

Phosphoric acid, 85% aq. soln., Thermo Scientific Chemicals

Catalog number: A18067.0D
10,000 mL, Each
Quantity
Catalog number: A18067.0D
also known as A18067-0D
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-38-2
IUPAC Name
phosphoric acid
Molecular Formula
H3O4P
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES
OP(O)(O)=O
Appearance (Color)
Clear colorless
Form
Viscous liquid or low-melting solid
Assay (Aqueous acid-base Titration)
84.0-87.0% (non-U.S. specification)
Assay from Suppliers CofA
84.0-87.0% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It is also used in dentistry and orthodontics as an etching solution as well as a sanitizing agent in dairy, food and brewing industries. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion in to water soluble phosphate compound.

Notes
Incompatible with strong bases and powdered metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text