Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
Catalog number A18070.0B
also known as A18070-0B
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
Add to cart
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Catalog numberA18070.0B
Price (USD)/ Each
68.65
Online exclusive
75.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS56378-72-4
IUPAC Namepentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
Molecular FormulaC4H12Mg5O19
InChI KeySXKPPTNZIUYFJR-UHFFFAOYSA-D
SMILESO.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Complexometric Titration≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White
FormPowder
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water and acids. Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.Rigolo; R.T.Woodhams. Basic magnesium carbonate flame retardants for polypropylene. Polymer Engineering & Science. 1992, 32,(5), 327-334.
  2. Supon Ananta; Rik Brydson; Noel W.Thomas. Synthesis, formation and characterisation of MgNb2O6 powder in a columbite-like phase. Journal of the European Ceramic Society. 1999, 19,(3), 355-362 .