DL-Methionine sulfoxide is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
Solubility
Soluble in water and Hcl.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
RUO – Research Use Only
General References:
- Bart Ghesquière; Veronique Jonckheere; Niklaas Colaert; Joost Van Durme; Evy Timmerman; Marc Goethals; Joost Schymkowitz; Frederic Rousseau; Joël Vandekerckhove; Kris Gevaert. Redox proteomics of protein-bound methionine oxidation. Molecular & Cellular Proteomics.2011, 10, (5), M110-006866.