Thermo Scientific Chemicals

DL-Methionine sulfoxide, 98+%

Catalog number: A18081.14
25 g, Each
Thermo Scientific Chemicals

DL-Methionine sulfoxide, 98+%

Catalog number: A18081.14
25 g, Each
Quantity
Catalog number: A18081.14
also known as A18081-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62697-73-8
IUPAC Name
2-amino-4-methanesulfinylbutanoic acid
Molecular Formula
C5H11NO3S
InChI Key
QEFRNWWLZKMPFJ-UHFFFAOYNA-N
SMILES
CS(=O)CCC(N)C(O)=O
Appearance (Color)
White
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (HPLC)
≥98.0%

Description

DL-Methionine sulfoxide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.

Solubility
Soluble in water and Hcl.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text