Hydrocortisone acetate, 97+%, Thermo Scientific Chemicals
Hydrocortisone acetate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrocortisone acetate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A18089.03
also known as A18089-03
Price (USD)/ Each
43.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
43.10
Add to cart
Hydrocortisone acetate, 97+%, Thermo Scientific Chemicals
Catalog numberA18089.03
Price (USD)/ Each
43.10
-
Add to cart
Chemical Identifiers
CAS50-03-3
SpecificationsSpecification SheetSpecification Sheet
CommentPurchased in the U.S. and in other countries
Melting Point (clear melt)214.0-224.0°C (non-U.S. specifiaction)
Optical Rotation+158° to +166° (c=1 in dioxane) (non-U.S. specification)
Appearance (Color)White to cream
Assay from Suppliers CofA≥97.0% (U.S. specification)
View more
Hydrocortisone acetate is used to treat hemorrhoids and itching/swelling in the rectum and anus. It is used to treat certain intestinal problems (such as ulcerative colitis of the rectum and other rectal/anal inflammatory conditions).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.
RUO – Research Use Only
Dan-Ni Wang; Takahito Toyotome; Yasunori Muraosa; Akira Watanabe; Tuya Wuren; Somnuk Bunsupa; Kaori Aoyagi; Mami Yamazaki; Masahiko Takino; Katsuhiko Kamei. GliA in Aspergillus fumigatus is required for its tolerance to gliotoxin and affects the amount of extracellular and intracellular gliotoxin. Medical Mycology.2014, 52, (5), 506-518.