Thermo Scientific Chemicals

Hydrocortisone acetate, 97+%, Thermo Scientific Chemicals

Catalog number: A18089.06
5 g, Each
Thermo Scientific Chemicals

Hydrocortisone acetate, 97+%, Thermo Scientific Chemicals

Catalog number: A18089.06
5 g, Each
Quantity
Catalog number: A18089.06
also known as A18089-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-03-3
Comment
Purchased in the U.S. and in other countries
Melting Point (clear melt)
214.0-224.0°C (non-U.S. specifiaction)
Optical Rotation
+158° to +166° (c=1 in dioxane) (non-U.S. specification)
Form
Powder or crystals or crystalline powder
Assay (HPLC)
≥97.0% (non-U.S. specification)

Description

Hydrocortisone acetate is used to treat hemorrhoids and itching/swelling in the rectum and anus. It is used to treat certain intestinal problems (such as ulcerative colitis of the rectum and other rectal/anal inflammatory conditions).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text