Thermo Scientific Chemicals

DL-Menthol, 98+%, Thermo Scientific Chemicals

Catalog number: A18098.0B
1000 g, Each
Thermo Scientific Chemicals

DL-Menthol, 98+%, Thermo Scientific Chemicals

Catalog number: A18098.0B
1000 g, Each
Quantity
Catalog number: A18098.0B
also known as A18098-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-78-1
IUPAC Name
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular Formula
C10H20O
InChI Key
NOOLISFMXDJSKH-KXUCPTDWSA-N
SMILES
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid

Description

Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its cooling action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat bad breath. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. Spud and Kool brands) for both its flavor and also to reduce irritation, especially to the throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, paraffin oil and water, 434.5 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text