DL-Menthol, 98+%, Thermo Scientific Chemicals
DL-Menthol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Menthol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
250 g
1000 g
Catalog number A18098.22
also known as A18098-22
Price (USD)/ Each
42.65
Online exclusive
47.50 
Save 4.85 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
47.50 
Save 4.85 (10%)
Add to cart
DL-Menthol, 98+%, Thermo Scientific Chemicals
Catalog numberA18098.22
Price (USD)/ Each
42.65
Online exclusive
47.50 
Save 4.85 (10%)
-
Add to cart
Chemical Identifiers
CAS89-78-1
IUPAC Name(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular FormulaC10H20O
InChI KeyNOOLISFMXDJSKH-KXUCPTDWSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its cooling action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat bad breath. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. Spud and Kool brands) for both its flavor and also to reduce irritation, especially to the throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, paraffin oil and water, 434.5 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. C. S. Sell. Sandhu. A Fragrant Introduction to Terpenoid Chemistry. Royal Society of Chemistry.;2003,76-77 and 83-93.
  2. Ma Carmen Romero.; Ma Concepción Navarro.; Joaquina Martín-Sánchez.; Adela Valero. Peppermint (Mentha piperita) and albendazole against anisakiasis in an animal model.Trop Med Int Health.,2014,19(21), 1430-1436.