It acts as an useful isomer of nicotinic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts as an useful isomer of nicotinic acid.
Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.
Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Guzman M.; Troncoso AM .; Guillen R .; Benitez M A. Isonicotinic acid: properties and analytical methods. Review.Boll. Chim. Farm.;1984,123200-209.
E. A. Zeller.; J. Barsky.; J. R. Fouts.; W. F. Kirchheimer.; L. S. Van Orden. Influence of isonicotinic acid hydrazide (INH) and 1-isonicotinyl-2-isopropyl hydrazide (IIH) on bacterial and mammalian enzymes.Experientia.,1952,8(9), 349-350.