Thermo Scientific Chemicals

3-Hydroxyflavone, 98+%, Thermo Scientific Chemicals

Catalog number: A18110.03
1 g, Each
Thermo Scientific Chemicals

3-Hydroxyflavone, 98+%, Thermo Scientific Chemicals

Catalog number: A18110.03
1 g, Each
Quantity
Catalog number: A18110.03
also known as A18110-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
577-85-5
Appearance (Color)
White to yellow or pale cream
Melting Point (clear melt)
168.0-174.0°C
Assay (HPLC)
≥98.0%
Form
Powder

Description

3-Hydroxyflavone is a reactant involved in studies of photochemically-induced dioxygenase-type CO-release reactivity; phase-transfer protection and deprotection of hydroxychromones; and O-methylation with di-Me carbonate. As a reactant it is involved in the synthesis of biologically active molecules including 2-chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors; dihydrochromenopyrazines and chromenoquinoxalines. It is also involved in studies of its electrochemical properties using voltammetric methodologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyflavone is a reactant involved in studies of photochemically-induced dioxygenase-type CO-release reactivity; phase-transfer protection and deprotection of hydroxychromones; and O-methylation with di-Me carbonate. As a reactant it is involved in the synthesis of biologically active molecules including 2-chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors; dihydrochromenopyrazines and chromenoquinoxalines. It is also involved in studies of its electrochemical properties using voltammetric methodologies.

Solubility
Insoluble in water. Soluble in N,N-DMF and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text