Thermo Scientific Chemicals

Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals

Catalog number: A18111.0F
2500 mL, Each
Thermo Scientific Chemicals

Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals

Catalog number: A18111.0F
2500 mL, Each
Quantity
Catalog number: A18111.0F
also known as A18111-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-27-0
IUPAC Name
propan-2-yl tetradecanoate
Molecular Formula
C17H34O2
InChI Key
AXISYYRBXTVTFY-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC(=O)OC(C)C
Appearance (Color)
Clear, colorless
Form
Liquid
Assay (GC)
>97.5%
Refractive Index
1.4330-1.4370 @ 20?C

Description

Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

Solubility
Miscible with alcohol. Immiscible with water and glycerol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text