2-Benzoylnaphthalene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Benzoylnaphthalene is used as pharmaceutical intermediate.
Solubility
Soluble in hot methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- D.Bhattacharya; S.Sharma; A.P.Singh. Selective benzoylation of naphthalene to 2-benzoylnaphthalene using zeolite H-beta catalysts. Applied Catalysis A: General. 1997, 150,(1), 53-62.
- John R.Ebdon; Ian Soutar; Paul Brown; Anthony R.Lane; Alasdair J.McCabe; Linda Swanson. Luminescence studies of polymer matrices: 2. on the phosphorescence characteristics of 2-benzoylnaphthalene dispersed in various acrylic polymers. High Performance Polymers. 1999, 11,(1), 49-62.