Tribromofluoromethane, 99%, stab. with copper, Thermo Scientific Chemicals
Tribromofluoromethane, 99%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tribromofluoromethane, 99%, stab. with copper, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
50 g
Catalog number A18124.06
also known as A18124-06
Price (USD)/ Each
79.65
Online exclusive
88.40 
Save 8.75 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.40 
Save 8.75 (10%)
Add to cart
Tribromofluoromethane, 99%, stab. with copper, Thermo Scientific Chemicals
Catalog numberA18124.06
Price (USD)/ Each
79.65
Online exclusive
88.40 
Save 8.75 (10%)
-
Add to cart
Chemical Identifiers
CAS353-54-8
IUPAC Nametribromo(fluoro)methane
Molecular FormulaCBr3F
InChI KeyIHZAEIHJPNTART-UHFFFAOYSA-N
SMILESFC(Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.5%
Appearance (Color)Clear, colorless to pale yellow
FormLiquid
Refractive Index1.5240-1.5270 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only