Thermo Scientific Chemicals

Tribromofluoromethane, 99%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A18124.14
25 g, Each
Thermo Scientific Chemicals

Tribromofluoromethane, 99%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A18124.14
25 g, Each
Quantity
Catalog number: A18124.14
also known as A18124-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
353-54-8
IUPAC Name
tribromo(fluoro)methane
Molecular Formula
CBr3F
InChI Key
IHZAEIHJPNTART-UHFFFAOYSA-N
SMILES
FC(Br)(Br)Br
Assay (GC)
>98.5%
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Refractive Index
1.5240-1.5270 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text