2-Chlorothioxanthone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorothioxanthone is used as pharmaceutical intermediate.
Solubility
Partly miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
R.B.Cundall; R.K.Chowdhary; A.K.Davies; V.P.Russell. Oxidation photosensitized by 2-chlorothioxanthone: the role of singlet oxygen. Journal of Photochemistry. 1984, 25,(2-4), 267-274.
Luis E.Pinero Santiago; Carmelo Garcia; Virginie Lhiaubet-Vallet; Miguel A.Miranda; Rolando Oyola. Solvent dependence of the photophysical properties of 2-chlorothioxanthone, the principal photoproduct of chlorprothixene. Photochemistry and Photobiology. 2011, 87,(3), 611-617.