Thermo Scientific Chemicals

Cacodylic acid sodium salt trihydrate 98+%, Thermo Scientific Chemicals

Catalog number: A18139.14
25 g, Each
Thermo Scientific Chemicals

Cacodylic acid sodium salt trihydrate 98+%, Thermo Scientific Chemicals

Catalog number: A18139.14
25 g, Each
Quantity
Catalog number: A18139.14
also known as A18139-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6131-99-3
IUPAC Name
sodium dimethylarsinate
Molecular Formula
C2H6AsNaO2
InChI Key
IHQKEDIOMGYHEB-UHFFFAOYSA-M
SMILES
[Na+].C[As](C)([O-])=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.

Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.

Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text