4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.
Solubility
Soluble in water (0.5 g/L).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
General References:
- Margaret Jefraim Jorgenson; Donald R.Hartter. A critical re-evaluation of the hammett acidity function at moderate and high acid concentrations of sulfuric acid. New H0 values based solely on a set of primary aniline indicators. J. Am. Chem. Soc. 1963, 85,(7), 878-883.
- Lingyan Zhu; Chiew Boon Tay; Hian Kee Lee. Liquid-liquid-liquid microextraction of aromatic amines from water samples combined with high-performance liquid chromatography. Journal of Chromatography A. 2002, 963,(1-2), 231-237.