It is employed as an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an intermediate in organic syntheses.
Solubility
Soluble in methanol (25 mg/ml).
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Hilton ST, et al. A tandem radical approach to the ABCE-rings of the Aspidosperma and Strychnos alkaloids.Chem. Commun. (Camb.) 2,2001,209-210.