L-Lysine methyl ester dihydrochloride, 99%
L-Lysine methyl ester dihydrochloride, 99%
Thermo Scientific Chemicals

L-Lysine methyl ester dihydrochloride, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A18157.22
also known as A18157-22
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
Add to cart
L-Lysine methyl ester dihydrochloride, 99%
Catalog numberA18157.22
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS26348-70-9
IUPAC Namedihydrogen methyl (2S)-2,6-diaminohexanoate dichloride
Molecular FormulaC7H18Cl2N2O2
InChI KeySXZCBVCQHOJXDR-ILKKLZGPSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Optical Rotation+18° to +25° (c=1 in methanol)
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ying Zhang; Ren-xi Zhuo. Synthesis and in vitro drug release behavior of amphiphilic triblock copolymer nanoparticles based on poly (ethylene glycol) and polycaprolactone. Biomaterials. 2005, 26,(33), 6736-6742.
  2. Andrew L.Lynch; Rongjun Chen; Paul J.Dominowski; Evgenyi Y.Shalaev; Robert J.Yancey Jr.; Nigel K.H.Slater. Biopolymer mediated trehalose uptake for enhanced erythrocyte cryosurvival. Biomaterials. 2010, 31,(23), 6096-6103.