Thermo Scientific Chemicals

DL-Ornithine monohydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A18173.14
25 g, Each
Thermo Scientific Chemicals

DL-Ornithine monohydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A18173.14
25 g, Each
Quantity
Catalog number: A18173.14
also known as A18173-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1069-31-4
IUPAC Name
hydrogen 2,5-diaminopentanoic acid chloride
Molecular Formula
C5H13ClN2O2
InChI Key
GGTYBZJRPHEQDG-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NCCCC(N)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%

Description

Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text