Thermo Scientific Chemicals

Zinc chromate, Thermo Scientific Chemicals

Catalog number: A18178.0B
1000 g, Each
Thermo Scientific Chemicals

Zinc chromate, Thermo Scientific Chemicals

Catalog number: A18178.0B
1000 g, Each
Quantity
Catalog number: A18178.0B
also known as A18178-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13530-65-9
Appearance (Color)
Yellow to orange-yellow
Form
crystalline powder
Elemental Analysis
Cr: 25.80 - 31.53% (theory 28.67%)
Elemental Analysis
Zn: 32.45 - 39.65% (theory 36.05%)

Description

Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text