Chemical Identifiers
CAS2645-08-1
IUPAC Namehydrogen ethyl 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoate chloride
Molecular FormulaC15H23ClN4O3
InChI KeyHIXDELXKSSLIKB-UHFFFAOYNA-N
SMILES[H+].[Cl-].CCOC(=O)C(CCCN=C(N)N)NC(=O)C1=CC=CC=C1
View more