Thermo Scientific Chemicals

Isonicotinamide, 99%, Thermo Scientific Chemicals

Catalog number: A18190.22
100 g, Each
Thermo Scientific Chemicals

Isonicotinamide, 99%, Thermo Scientific Chemicals

Catalog number: A18190.22
100 g, Each
Quantity
Catalog number: A18190.22
also known as A18190-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1453-82-3
IUPAC Name
pyridine-4-carboxamide
Molecular Formula
C6H6N2O
InChI Key
VFQXVTODMYMSMJ-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=NC=C1
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
153.0-160.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text