Thermo Scientific Chemicals

3-Cyclopentylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18192.14
25 g, Each
Thermo Scientific Chemicals

3-Cyclopentylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18192.14
25 g, Each
Quantity
Catalog number: A18192.14
also known as A18192-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-77-2
IUPAC Name
3-cyclopentylpropanoic acid
Molecular Formula
C8H14O2
InChI Key
ZRPLANDPDWYOMZ-UHFFFAOYSA-N
SMILES
OC(=O)CCC1CCCC1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Refractive Index
1.4555-1.4595 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text