Thermo Scientific Chemicals

2-Amino-6-chloropurine, 99%, Thermo Scientific Chemicals

Catalog number: A18195.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-6-chloropurine, 99%, Thermo Scientific Chemicals

Catalog number: A18195.14
25 g, Each
Quantity
Catalog number: A18195.14
also known as A18195-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10310-21-1
IUPAC Name
6-chloro-7H-purin-2-amine
Molecular Formula
C5H4ClN5
InChI Key
RYYIULNRIVUMTQ-UHFFFAOYSA-N
SMILES
NC1=NC(Cl)=C2NC=NC2=N1
Water Content (Karl Fischer Titration)
<0.5%
Assay (HPLC)
>98.5%
Appearance (Color)
White to cream or pale yellow
Form
Powder
Assay (Non-aqueous acid-base Titration)
>98.5 to <101.5% (dry wt. basis)

Description

2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2?-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2′-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.

Solubility
Solubility in 1mol/L NaOH; very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text