N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific Chemicals
N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A18199.14
also known as A18199-14
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
Add to cart
N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific Chemicals
Catalog numberA18199.14
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS3088-42-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only