Thermo Scientific Chemicals

4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A18200.14
25 g, Each
Thermo Scientific Chemicals

4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A18200.14
25 g, Each
Quantity
Catalog number: A18200.14
also known as A18200-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16004-15-2
IUPAC Name
1-(bromomethyl)-4-iodobenzene
Molecular Formula
C7H6BrI
InChI Key
BQTRMYJYYNQQGK-UHFFFAOYSA-N
SMILES
BrCC1=CC=C(I)C=C1
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
75.0-84.0?C
Assay (GC)
≥96.0%

Description

4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text