Thermo Scientific Chemicals

L-(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18203.14
25 g, Each
Thermo Scientific Chemicals

L-(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18203.14
25 g, Each
Quantity
Catalog number: A18203.14
also known as A18203-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20312-36-1
IUPAC Name
(2S)-2-hydroxy-3-phenylpropanoate
Molecular Formula
C9H9O3
InChI Key
VOXXWSYKYCBWHO-QMMMGPOBSA-M
SMILES
O[C@@H](CC1=CC=CC=C1)C([O-])=O
Appearance (Color)
White to pale cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
121.0-127.0?C
Optical Rotation
-22.0 ± 2.0? (C = 1 in water)

Description

L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.

Solubility
Solubility in water gives very faint turbidity. 

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text