1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only
Lukas Meier; Stefan Schmid; Christian Berchtold; Renato Zenobi. Contribution of liquid-phase and gas-phase ionization in extractive electrospray ionization mass spectrometry of primary amines. European Journal of Mass Spectrometry. 2011, 17,(4), 345-351.
T.N.Guru Row. Hydrogen and fluorine in crystal engineering: systematics from crystallographic studies of hydrogen bonded tartrate-amine complexes and fluoro-substituted coumarins, styrylcoumarins and butadienes. Coordination Chemistry Reviews. 1999, 183,(1), 81-100.