Thermo Scientific Chemicals

n-Tridecane, 98+%, Thermo Scientific Chemicals

Catalog number: A18220.36
500 g, Each
Thermo Scientific Chemicals

n-Tridecane, 98+%, Thermo Scientific Chemicals

Catalog number: A18220.36
500 g, Each
Quantity
Catalog number: A18220.36
also known as A18220-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-50-5
IUPAC Name
tridecane
Molecular Formula
C13H28
InChI Key
IIYFAKIEWZDVMP-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCC
Form
Liquid
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
≥98.0%

Description

n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text