Contains mono-, tri- and tetrabromofluorescein, average composition is dibromo.
Comment
Melting point varies with balance of mono-, tri- and tetrabromo- forms.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.