Thermo Scientific Chemicals

Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals

Catalog number: A18231.22
100 g, Each
Thermo Scientific Chemicals

Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals

Catalog number: A18231.22
100 g, Each
Quantity
Catalog number: A18231.22
also known as A18231-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-43-3
IUPAC Name
1,10-dibutyl decanedioate
Molecular Formula
C18H34O4
InChI Key
PYGXAGIECVVIOZ-UHFFFAOYSA-N
SMILES
CCCCOC(=O)CCCCCCCCC(=O)OCCCC
Assay (GC)
≥90.0%
Refractive Index
1.4375-1.4445 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text