Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals
Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number A18231.36
also known as A18231-36
Price (USD)/ Each
111.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
111.00
Add to cart
Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals
Catalog numberA18231.36
Price (USD)/ Each
111.00
-
Add to cart
Chemical Identifiers
CAS109-43-3
IUPAC Name1,10-dibutyl decanedioate
Molecular FormulaC18H34O4
InChI KeyPYGXAGIECVVIOZ-UHFFFAOYSA-N
SMILESCCCCOC(=O)CCCCCCCCC(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥90.0%
Refractive Index1.4375-1.4445 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only