Thermo Scientific Chemicals

1-Hexanol, 99%, Thermo Scientific Chemicals

Catalog number: A18232.0F
2500 mL, Each
Thermo Scientific Chemicals

1-Hexanol, 99%, Thermo Scientific Chemicals

Catalog number: A18232.0F
2500 mL, Each
Quantity
Catalog number: A18232.0F
also known as A18232-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-27-3
IUPAC Name
hexan-1-ol
Molecular Formula
C6H14O
InChI Key
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILES
CCCCCCO
Identification (FTIR)
Conforms (UK Sourced Material)
Comment
Material Sourced in UK and US
Form
Liquid
Assay (GC)
≥98.5% (UK Source Material)
Assay from Supplier's CofA
≥98.5% (US Sourced Material)

Description

1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.

Solubility
Miscible with ethanol, acetone, chloroform, ether, benzene. Slightly miscible with carbon tetrachloride and water.

Notes
Incompatible with strong acids, acid chlorides, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text