Thermo Scientific Chemicals

2,2,2-Trifluoroethyl acrylate, 98%, stab. with 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: A18235.06
5 g, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl acrylate, 98%, stab. with 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: A18235.06
5 g, Each
Quantity
Catalog number: A18235.06
also known as A18235-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
407-47-6
Refractive Index
1.3480-1.3520 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Solution Test
Clear (10% in Diethyl ether, non-U.S. specification)

Description

It is used for adhesives because of its peeling property and also used for contact lens to give antifouling property. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for adhesives because of its peeling property and also used for contact lens to give antifouling property. It is also employed as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, heat, light UV radiation and free radical initiators. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light and electrostatic charges. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text