2-Chlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2-Chlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A18238.14
also known as A18238-14
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
2-Chlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog numberA18238.14
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS2740-81-0
IUPAC Name1-chloro-2-isothiocyanatobenzene
Molecular FormulaC7H4ClNS
InChI KeyDASSPOJBUMBXLU-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.6595-1.6645 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only