N(alpha)-Benzyloxycarbonyl-L-glutamine, 99%, Thermo Scientific Chemicals
N(alpha)-Benzyloxycarbonyl-L-glutamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N(alpha)-Benzyloxycarbonyl-L-glutamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18240.06
also known as A18240-06
Price (USD)/ Each
24.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
24.90
Add to cart
N(alpha)-Benzyloxycarbonyl-L-glutamine, 99%, Thermo Scientific Chemicals
Catalog numberA18240.06
Price (USD)/ Each
24.90
-
Add to cart
Chemical Identifiers
CAS2650-64-8
IUPAC Name(2S)-2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanoic acid
Molecular FormulaC13H16N2O5
InChI KeyJIMLDJNLXLMGLX-JTQLQIEISA-N
SMILESNC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)133.0-139.0?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation-7.1? ± 0.5? (c=2 in ethanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only