Thermo Scientific Chemicals

1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A18255.14
25 g, Each
Thermo Scientific Chemicals

1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A18255.14
25 g, Each
Quantity
Catalog number: A18255.14
also known as A18255-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-41-8
IUPAC Name
1-chloro-2-iodobenzene
Molecular Formula
C6H4ClI
InChI Key
MPEOPBCQHNWNFB-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1I
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Refractive Index
1.6335-1.6365 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid

Description

1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2'-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2′-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place. Keep the container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text