N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A18257.22
also known as A18257-22
Price (USD)/ Each
76.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
76.70
Add to cart
N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
Catalog numberA18257.22
Price (USD)/ Each
76.70
-
Add to cart
Chemical Identifiers
CAS1888-91-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4875-1.4905 @ 20?C
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only