Thermo Scientific Chemicals

Potassium bromate, 99%, Thermo Scientific Chemicals

Catalog number: A18258.36
500 g, Each
Thermo Scientific Chemicals

Potassium bromate, 99%, Thermo Scientific Chemicals

Catalog number: A18258.36
500 g, Each
Quantity
Catalog number: A18258.36
also known as A18258-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-01-2
IUPAC Name
potassium bromate
Molecular Formula
BrKO3
InChI Key
OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES
[K+].[O-][Br](=O)=O
Assay (Redox Titration)
>98.5 to <101.5%
Form
Crystalline powder
Appearance (Color)
White

Description

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.

Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text