Thermo Scientific Chemicals

meso-Tetraphenylporphine, low chlorine, Thermo Scientific Chemicals

Catalog number: A18261.06
5 g, Each
Thermo Scientific Chemicals

meso-Tetraphenylporphine, low chlorine, Thermo Scientific Chemicals

Catalog number: A18261.06
5 g, Each
Quantity
Catalog number: A18261.06
also known as A18261-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
917-23-7
Appearance (Color)
Dark blue to dark purple
Elemental Analysis
H: 4.62 - 5.22% (Theoretical 4.92%)
Elemental Analysis
C: 85.11 - 86.26% (Theoretical 85.97%)
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder

Description

meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text