Thermo Scientific Chemicals

Calcium 2-ethylhexanoate, 98%, Thermo Scientific Chemicals

Catalog number: A18266.14
25 g, Each
Thermo Scientific Chemicals

Calcium 2-ethylhexanoate, 98%, Thermo Scientific Chemicals

Catalog number: A18266.14
25 g, Each
Quantity
Catalog number: A18266.14
also known as A18266-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
136-51-6
IUPAC Name
calcium bis(2-ethylhexanoate)
Molecular Formula
C16H30CaO4
InChI Key
LTPCXXMGKDQPAO-UHFFFAOYNA-L
SMILES
[Ca++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Appearance (Color)
White to pale cream
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Calcium 2-ethylhexanoate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium 2-ethylhexanoate is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text