Thermo Scientific Chemicals

Naphthol Green B

Catalog number: A18268.22
100 g, Each
Thermo Scientific Chemicals

Naphthol Green B

Catalog number: A18268.22
100 g, Each
Quantity
Catalog number: A18268.22
also known as A18268-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19381-50-1
IUPAC Name
trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron
Molecular Formula
C30H18FeN3Na3O15S3
InChI Key
VLBLPLXOYXEXJK-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
UV/Vis
Conforms
Appearance (Color)
Dark green to black

Description

It finds its applications as a leather and fur dye and as a biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its applications as a leather and fur dye and as a biological stain.

Solubility
Soluble in water (160 mg/ml at 20°C), ethanol (0.9 mg/ml), and ethylene (9 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text