Thermo Scientific Chemicals

Scandium(III) acetate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: A18271.03
1 g, Each
Thermo Scientific Chemicals

Scandium(III) acetate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: A18271.03
1 g, Each
Quantity
Catalog number: A18271.03
also known as A18271-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
304675-64-7
IUPAC Name
scandium(3+) triacetate
Molecular Formula
C6H9O6Sc
InChI Key
DBTMQFKUVICLQN-UHFFFAOYSA-K
SMILES
[Sc+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Appearance (Color)
White
Total Metal Impurities
≤0.1%
Assay (unspecified)
≥99.9%
Form
Powder

Description

Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text