Thermo Scientific Chemicals

Phenyl propargyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: A18272.03
1 g, Each
Thermo Scientific Chemicals

Phenyl propargyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: A18272.03
1 g, Each
Quantity
Catalog number: A18272.03
also known as A18272-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13610-02-1
Refractive Index
1.5315-1.5355 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥98.0%

Description

Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water (1.0 g/L at 25°C).

Notes
Air and light sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text