Thermo Scientific Chemicals

2,3,5,6-Tetramethylpyrazine, 98+%, Thermo Scientific Chemicals

Catalog number: A18279.22
100 g, Each
Thermo Scientific Chemicals

2,3,5,6-Tetramethylpyrazine, 98+%, Thermo Scientific Chemicals

Catalog number: A18279.22
100 g, Each
Quantity
Catalog number: A18279.22
also known as A18279-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1124-11-4
IUPAC Name
tetramethylpyrazine
Molecular Formula
C8H12N2
InChI Key
FINHMKGKINIASC-UHFFFAOYSA-N
SMILES
CC1=NC(C)=C(C)N=C1C
Assay (GC)
≥98.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Melting Point
83.5-89.5?C

Description

2,3,5,6-Tetramethylpyrazine is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5,6-Tetramethylpyrazine is a flavor ingredient of tobacco. It forms cocrystal with 4-hydroxybenzoic acid which exhibits supramolecular synthon polymorphism.

Solubility
Soluble in water (4 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text